Palladium copper on alumina (1% Pd, 0.3% Cu), granules, reduced
44872-14
44872-14
AAA44872-14EA
167.61
USD
AAA44872-14
CAAAA44872-14
Palladium copper on alumina (1% Pd, 0.3% Cu), granules, reduced
Palladium copper on alumina
Granular
Storage Temperature:
Ambient |
MDL Number:
MFCD08064572 CAS Number: 7440-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...