Palladium copper on alumina (1% Pd, 0.3% Cu), spheres 2 mm, reduced

44833-14 44833-22 44833-36 44833-36 44833-14 44833-22
AAA44833-14EA 225.15 USD
AAA44833-14 AAA44833-36 AAA44833-22 CAAAA44833-22 CAAAA44833-36 CAAAA44833-14
Palladium copper on alumina (1% Pd, 0.3% Cu), spheres 2 mm, reduced
Palladium copper on alumina
Storage Temperature: Ambient
MDL Number: MFCD08064572
CAS Number: 7440-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR