Nafion® R-1100 resin, sulfonyl fluoride form

Supplier: Alfa Aesar
Warning

44626-09 44626-04 44626-30 44626-30 44626-09 44626-18 44626-04 44626-18
AAA44626-09EA 159.03 USD
AAA44626-09 AAA44626-04 CAAAA44626-30 AAA44626-30 CAAAA44626-09 CAAAA44626-18 CAAAA44626-04 AAA44626-18
Nafion® R-1100 resin, sulfonyl fluoride form
Nafion® sulfonyl fluoride form

Pellets


Applications: Thermoplastic-can be extruded or laminated

Formula: C₉F₁₈O₄S
MW: 546.126 g/mol
Melting Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD05665455
CAS Number: 26654-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR