Cyclohexyl methyl methylphosphonate 99%

Supplier: Thermo Scientific Chemicals
Warning

44426.04 44426-09 44426-04 44426-02 44426.02
AAA44426-09EA 952.75 USD
AAA44426-09 AAA44426-04 AAA44426-02 CAAAA44426-02 CAAAA44426-04
Cyclohexyl methyl methylphosphonate 99%
Cyclohexyl methyl methylphosphonate
Formula: C₈H₁₇O₃P
Storage Temperature: Ambient
MDL Number: MFCD08064211
CAS Number: 7040-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR