Tris[N,N-bis(trimethylsilyl)amide]cerium(III) 98%

Supplier: Thermo Scientific Chemicals
Danger

44362.14 44362-14
AAA44362-14EA 1813.99 USD
CAAAA44362-14 AAA44362-14
Tris[N,N-bis(trimethylsilyl)amide]cerium(III) 98%
Tris[N,N-bis(trimethylsilyl)amide]cerium (III)

Powder

Formula: C₁₈H₅₄CeN₃Si₆
MW: 621.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411241
CAS Number: 41836-21-9
UN: 3396
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR