Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1S)-(+)-Camphor-10-sulfonamide 97%

Supplier: Alfa Aesar

44361-14 44361-14 44361-03 44361-03 44361-06 44361-06
AAA44361-14EA 491.31 USD InStock
AAA44361-14 CAAAA44361-14 CAAAA44361-03 AAA44361-03 AAA44361-06 CAAAA44361-06
(1S)-(+)-Camphor-10-sulfonamide 97%
(1S)-(+)-Camphor-10-sulphonamide
Powder


Formula: C₁₀H₁₇NO₃S
MW: 231,32
Melting Pt: 133…135 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075611
CAS Number: 60933-63-3
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty