Bis(cyclopentadienyl)vanadium bromide 98%

Danger


ampouled under argon

44360-06 44360-03 44360-06 44360-03
AAA44360-06EA 266.4 USD
CAAAA44360-03 AAA44360-06 AAA44360-03 CAAAA44360-06
Bis(cyclopentadienyl)vanadium bromide 98%
Bis(cyclopentadienyl)vanadium bromide
Formula: C₁₀H₁₀BrV
Boiling Pt: 165 °C (0.2 mmHg) (subl.)
Melting Pt: 221…222 °C
Storage Temperature: Ambient
MDL Number: MFCD03701567
CAS Number: 64815-29-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR