(1R)-(-)-Camphor-10-sulfonamide 97%

44358-14 44358-06 44358-03 44358-03 44358-06
AAA44358-06EA 130.41 USD
CAAAA44358-03 AAA44358-06 CAAAA44358-06 AAA44358-14 AAA44358-03
(1R)-(-)-Camphor-10-sulfonamide 97%
(1R)-(-)-Camphor-10-sulphonamide

Powder

Formula: C₁₀H₁₇NO₃S
Melting Pt: 134 °C. (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151500
CAS Number: 72597-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR