Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 6.0 micron, fluorescent

Supplier: Alfa Aesar

44144-9W 44144-9W
AAA44144-9WEA 303.95 USD InStock
CAAAA44144-9W AAA44144-9W
Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 6.0 micron, fluorescent
Poly(styrene)
Applications: Diagnostic and protein binding assays


Formula: (C₈H₈)n
MW: 125,000-250,000
Storage Temperature: Refrigerator
MDL Number: MFCD00084450
CAS Number: 9003-53-6
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty