Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ruthenium(III) 99%

Warning

Synonyms: Ru(TMHD)3Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ruthenium(III)

44006-04 44006-02 44006-02 44006-04
AAA44006-02EA 146.95 USD
CAAAA44006-04 AAA44006-02 CAAAA44006-02 AAA44006-04
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ruthenium(III) 99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ruthenium(III)

Powder

Formula: C₃₃H₅₇O₆Ru
MW: 650,872 g/mol
Boiling Pt: 250°C dec,
Melting Pt: 200-203°C
Storage Temperature: Ambient
CAS Number: 38625-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR