Bismuth(III) isopropoxide
Supplier: Thermo Scientific Chemicals
|
Warning
|
43938.06
43938-06
AAA43938-06EA
532.99
USD
CAAAA43938-06
AAA43938-06
Bismuth(III) isopropoxide
Bismuth (III) isopropoxide
Powder, Packaged under argon
Formula:
C₉H₂₁BiO₃ MW: 386.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00799065 CAS Number: 15049-67-9 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...