2,1,3-Benzothiadiazole 98%

Supplier: Thermo Scientific Chemicals

43730-04 43730-09 43730-04 43730-09
AAA43730-09EA 62.6 USD
AAA43730-09 CAAAA43730-09 CAAAA43730-04 AAA43730-04
2,1,3-Benzothiadiazole 98%
2,1,3-Benzothiadiazole

Powder

Formula: C₆H₄N₂S
Boiling Pt: 206 °C
Melting Pt: 42…44 °C
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005809
CAS Number: 273-13-2
EINECS: 205-985-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR