Polyethyleneimine, on silica beads 40-200 mesh anion exchange resin, benzylated
Synonyms:
Polyethylenimine
, PEI
, Corcat
, Polymin P
, Ethyleneimine Polymer
, Aziridine
, Poly(ethyleneimine)
43305-AK
43305-AD
43305-AB
43305-AD
43305-AK
43305-AB
AAA43305-ABEA
29.59
USD
AAA43305-AB
CAAAA43305-AB
CAAAA43305-AD
AAA43305-AK
CAAAA43305-AK
AAA43305-AD
Polyethyleneimine, on silica beads 40-200 mesh anion exchange resin, benzylated
Polyethyleneimine
Powder
Applications: Solid phase organic synthesis, affinity chromatography, catalyst support
Formula:
(CH₂CH₂NH)n Storage Temperature: Ambient |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...