Ytterbium (III) trifluoromethanesulfonate hydrate 98%

Warning

Synonyms: Trifluoromethanesulfonic acid ytterbium salt hydrateTrifluoromethanesulphonic acid ytterbium salt hydrateYtterbium(III) trifluoromethanesulfonate hydrateYtterbium(III) triflate hydrateYtterbium triflate hydrateYtterbium(III) trifluoromethanesulphonate hydrate

40314-06 40314-06
AAA40314-06EA 79.26 USD
CAAAA40314-06 AAA40314-06
Ytterbium (III) trifluoromethanesulfonate hydrate 98%
Ytterbium (III) trifluoromethanesulphonate hydrate

Powder

Formula: C₃F₉O₉S₃Yb·xH₂O
MW: 620.25 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD03703499
CAS Number: 252976-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR