KLEENGUARD® A40 Liquid and Particle Protection Pant, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
KLEENGUARD®
44414 44415 44413
82007-652CS 243.21 USD
82007-652 82007-650 82007-648
KLEENGUARD® A40 Liquid and Particle Protection Pant, KIMBERLY-CLARK PROFESSIONAL®
Disposable Garments
KLEENGUARD® A40 Pants are designed to be used in applications where Tyvek® garments are typically worn, as they provide an excellent particle barrier and resist nonhazardous liquids
Meets ANSI/ISEA 101-1996. NFPA 99 compliant antistatic material. Passes ASTM F1670 testing for penetration of blood and bodily fluids.
Pants provide a better liquid barrier, better particulate barrier, and are lower in lint than Tyvek®. Dry particle holdout is 100% between 0.3–0.5µm. Since the fabric is a lamination of microporous film and a spunbond nonwoven material, the tough film protects, while the soft nonwoven fabric provides comfort against the skin. All pants feature an elastic waist.