Would you like to log in again?
KLEENGUARD® A80 Chemical Permeation & Jet Liquid Protection Sleeve Protector, KIMBERLY-CLARK PROFESSIONAL®
Supplier: KIMBERLY CLARK CORP
KLEENGUARD®
45749 45749
93000-726CS 257.8 USD InStock
CA93000-726 93000-726
KLEENGUARD® A80 Chemical Permeation & Jet Liquid Protection Sleeve Protector, KIMBERLY-CLARK PROFESSIONAL®
Apparel Sleeve Protectors
The KLEENGUARD® A80 line of apparel is designed to provide excellent worker protection in chemical handling/mixing/cleanup, hazardous material handling, hazardous waste cleanup, industrial Hazmat, and petrochemical applications
Sleeves are made from a durable fabric with an exterior laminated with SARANEX® 23-P film and an interior that features a comfortable, cloth-like feel against the skin.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty