Eight flexible storage options including 3 full-extension drawers, 1 large bulk storage area, 6 shelves, 2 built in dispensers for gloves and wipers, a 2 compartment tool and document holder.
- Chemically and corrosion resistant polyethylene construction
- Adds space, function, and mobility
- Sliding door
- ABS drawers
- 3 full-extension drawers
- 1 large bulk storage area
- 6 side wing shelves (2 with dispensing slots)
- Lock all drawers with 1 key
- 2 handles for easy mobility
- 4 totally sealed bearing casters (3 inch)
- American made by TrippNT
- Total Replacement Lifetime Guarantee
Organizing cart made from food grade white high density polyethylene has four 3 inch industrial total locking casters
This cart features 3 full-extension drawers (pull all the way out to make accessing drawer contents easy) that are 12-3/4W x 3-5/8H x 15-7/8D". Head space clearance from bottom of drawer to bottom of drawer above is 4-1/2".
The Core DX creates storage for glassware, chemicals, pens, pipets, tools, tweezers, log books, documents, bottles and other supplies. The top side shelf is a built in dispenser for Kimwipes and gloves. Shelves snap in and out as needed. The bulk storage area is 12-3/4W x 14-1/8H x 16-3/8D".
Utilized in labs, clinics, hospitals, doctor's offices and industry to add extra work space, storage space and mobility. The Core DX improves efficiency and moves supplies to the work area instead of making multiple trips to set up an activity.
Clearance: 41/2", Drawer Head Space
Delivery information: Ships completely assembled