Oligo p(dT)12-18 Sodium Salt, Affymetrix

19817100UN 198175UN 1981725UN
101175-118EA 389.23 USD
101175-118 101171-084 101174-576
Oligo p(dT)12-18 Sodium Salt, Affymetrix
Form: Lyophilized.

  • This oligodeoxythymidylate mixture is synthesized using O-cyanoethyl phosphoramidite chemistry. After the removal of the protecting groups, the product is converted to the sodium salt and the excess salt removed by gel filtration.

Unit Definition: One A260 unit is that quantity of material which, when dissolved in 1.0ml of water and read in a spectrophotometer at 260 nm in a cuvette with a 1.0 cm light path, gives an absorbance of 1.0. One A260 unit is approximately equal to 30-35 μg of DNA.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR