Would you like to log in again?
DNA Ladder, 1 kb plus, Affymetrix/USB
Supplier: Thermo Fisher Scientific Chemicals Inc.
J76714-YCR
The 1kb Plus DNA Ladder is a set of 14 easy-to-remember band sizes for use in qualitative agarose gel analysis ranging in size from 12kb down to 100bp
6X DNA Loading Buffer (BXF) with bromophenol blue and xylene cyanol FF is included for convenience to prepare DNA samples for agarose gel electrophoresis.
Fragments: 12 kb; 10 kb; 8 kb; 6 kb; 4 kb; 3 kb; 2 kb; 1.5 kb; 1 kb; 500 bp; 400 bp; 300 bp; 200 bp; 100 bp.
To Use: Add 5μl ladder mix per gel lane.
Two high intensity reference bands of 4kb and 500bp are included. The ladder is supplied in ready-to-load form in gel loading buffer. Simply add the recommended amount (5μl) for highly resolved bands.
ORDER
Expand All / Collapse All
Size Storage Temperature Supplier No. VWR Catalog Number Unit Price Quantity
500microL Frozen J76714-YCR 101375-596 Each (500µl) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty