Luria Broth (LB), Ready-Made Powder, Thermo Scientific
Supplier: Thermo Fisher Scientific Chemicals Inc.
J75854-A1
101170-308EA 124.25 USD
101170-308
Luria Broth (LB), Ready-Made Powder, Thermo Scientific
Media for Microbiology Dehydrated Media for Microbiology
For the propagation of bacteria.
Contents:
Off-white free flowing powder. To use: Dissolve 25gm powder per liter of purified water. Autoclave.
pH(2.5%, H2O): 7.0 ± 0.2