Phenol, pH 8.0, Equilibrated, MB Grade, Affymetrix
J75829-AN J75829-AE
101172-024EA 129.6 USD
101172-024 101174-858
Phenol, pH 8.0, Equilibrated, MB Grade, Affymetrix
Used in nucleic acid purification procedures and dissolution of protein.
This product is supplied as water-saturated phenol with a separate bottle of Phenol Equilibration Buffer. At the time of product use, simply add the entire contents of Phenol Equilibration Buffer to the water-saturated phenol.
Contaminant Testing: DNase: Passes test.
pH (5%, after equilibration with 75833, 20°C) after addition of Phenol Equilibration Buffer: ≥ 8.0