Betaine solution 5 M, ultrapure Molecular biology grade

Supplier: Thermo Fisher Scientific Chemicals Inc.

J77507-UCR J77507-AB
101174-140EA 29.88 USD InStock
101174-140 101375-612
Betaine solution 5 M, ultrapure Molecular biology grade
Betaine

5M solution of betaine in deionized dH2O. Functionally tested for use in PCR.


  • Contaminant Testing:
  • DNase (endo): Passes test
  • DNase (exo): Passes test
  • RNase: Passes test

Formula: C₅H₁₁NO₂
MW: 117,15
Storage Temperature: Freezer
MDL Number: MFCD00012123
CAS Number: 107-43-7
EINECS: 203-490-6

Order Now

Specification Test Results

DNase (endo) Passes test
DNase (exo) Passes test
RNase Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR