p-Phenylenediacetonitrile ≥98.0%

Supplier: TCI America

Synonyms: 1,4-Phenylenediacetonitrile2,2'-(1,4-Phenylene)diacetonitrile

X0061-25G X0061-5G
TCX0061-25GEA 308 USD
TCX0061-25G TCX0061-5G
p-Phenylenediacetonitrile ≥98.0%
p-Phenylenediacetonitrile

CAS Number: 622-75-3
MDL Number: MFCD00001923
Molecular Formula: C10H8N2
Molecular Weight: 156.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 98


Other Product Names / Synonyms: alpha,alpha'-Dicyano-p-xylene, 1,4-Phenylenediacetonitrile, p-Xylylene Cyanide

Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Melting Pt: 98 °C
MDL Number: MFCD00001923
CAS Number: 622-75-3
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/74/09/28997409.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR