Xanthopterin hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Amino-4,6-pteridinediol

X0007-1G X0007-5G
TCX0007-1GEA 107 USD
TCX0007-1G TCX0007-5G
Xanthopterin hydrate ≥98.0% (by HPLC, titration analysis)
Xanthopterin hydrate

CAS Number: 5979-01-1
MDL Number: MFCD00151241
Molecular Formula: C6H5N5O2
Molecular Weight: 179.14
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: Pale Yellow


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 2-Amino-4,6-pteridinediol

Formula: C₆H₅N₅O₂
MDL Number: MFCD00151241
CAS Number: 5979-01-1
https://us.vwr.com/stibo/web/std.lang.all/73/71/28997371.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR