4-Acetoxy-3-methoxybenzaldehyde ≥98.0%

Supplier: TCI America

Synonyms: Vanillin acetate4-Formyl-2-methoxyphenyl acetate

V0104-25G
TCV0104-25GEA 42 USD
TCV0104-25G
4-Acetoxy-3-methoxybenzaldehyde ≥98.0%
4-Acetoxy-3-methoxybenzaldehyde

CAS Number: 881-68-5
MDL Number: MFCD00003362
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 77


Other Product Names / Synonyms: 4-Acetoxy-m-anisaldehyde, 4-Acetoxy-3-methoxybenzaldehyde

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 77 °C
MDL Number: MFCD00003362
CAS Number: 881-68-5
https://us.vwr.com/stibo/web/std.lang.all/73/21/28997321.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR