N-Vinylphthalimide ≥98.0%

Supplier: TCI America

V0065-25G V0065-5G
TCV0065-25GEA 433 USD
TCV0065-25G TCV0065-005G
N-Vinylphthalimide ≥98.0%
N-Vinylphthalimide

CAS Number: 3485-84-5
MDL Number: MFCD00078446
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 151
Melting point (°C): 83

Formula: C₁₀H₇NO₂
Boiling Pt: 151 °C
Melting Pt: 83 °C
MDL Number: MFCD00078446
CAS Number: 3485-84-5
https://us.vwr.com/stibo/web/std.lang.all/73/01/28997301.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR