Ethyl vanillate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 4-hydroxy-3-methoxybenzoate

V0051-25G
TCV0051-025GEA 121.7 USD
TCV0051-025G
Ethyl vanillate ≥98.0%
Ethyl vanillate

CAS Number: 617-05-0
MDL Number: MFCD00017269
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 293
Melting point (°C): 43


Other Product Names / Synonyms: Vanillic Acid Ethyl Ester, 4-Hydroxy-3-methoxybenzoic Acid Ethyl Ester, Ethyl 4-Hydroxy-3-methoxybenzoate

Formula: C₁₀H₁₂O₄
MW: 196.20 g/mol
Boiling Pt: 293 °C
Melting Pt: 43 °C
MDL Number: MFCD00017269
CAS Number: 617-05-0
https://us.vwr.com/stibo/web/std.lang.all/72/89/28997289.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR