Sodium 1-undecanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

U0031-25G
TCU0031-025GEA 208.19 USD
TCU0031-025G
Sodium 1-undecanesulfonate ≥98.0% (by titrimetric analysis)
Sodium 1-undecanesulphonate

CAS Number: 5838-34-6
MDL Number: MFCD00053627
Molecular Formula: C11H24O3S
Molecular Weight: 258.35
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: 1-Undecanesulfonic Acid Sodium Salt

https://us.vwr.com/stibo/web/std.lang.all/71/93/28997193.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR