2,3,4-Trifluorophenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
Dihydroxy(2,3,4-trifluorophenyl)borane
, 2,3,4-Trifluorobenzeneboronic acid
, (2,3,4-Trifluorophenyl)metaboric acid
T2574-5G
TCT2574-5GEA
36.52
USD
TCT2574-5G
2,3,4-Trifluorophenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
2,3,4-Trifluorophenylboronic acid
CAS Number: 226396-32-3
MDL Number: MFCD01863168
Molecular Formula: C6H4BF3O2
Molecular Weight: 175.90
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 235
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2,3,4-Trifluorobenzeneboronic Acid
Formula:
C₆H₄BF₃O₂ MW: 175.9 g/mol Melting Pt: 235 °C |
MDL Number:
MFCD01863168 CAS Number: 226396-32-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...