(S)-(+)-Tetrahydrofurfurylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-(+)-2-(Aminomethyl)tetrahydrofuran
, (S)-(Tetrahydrofuran-2-yl)methanamine
CAS Number: 7175-81-7
MDL Number: MFCD00085303
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 52
Flash Point (°C): 46
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: 15 deg (C=2, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-2-(Aminomethyl)tetrahydrofuran
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 52 °C Density: 0.98 (20 °C) Flash Pt: 46 °C |
MDL Number:
MFCD00085303 CAS Number: 7175-81-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...