4-(Trifluoromethoxy)benzenesulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

T2427-25G T2427-5G
TCT2427-5GEA 60 USD
TCT2427-5G TCT2427-25G
4-(Trifluoromethoxy)benzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
4-(Trifluoromethoxy)benzenesulphonamide

CAS Number: 1513-45-7
MDL Number: MFCD00798381
Molecular Formula: C7H6F3NO3S
Molecular Weight: 241.18
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 149

Formula: C₇H₆F₃NO₃S
MW: 241.19 g/mol
Melting Pt: 149 °C
MDL Number: MFCD00798381
CAS Number: 1513-45-7
https://us.vwr.com/stibo/web/std.lang.all/61/80/28996180.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR