Tribromofluoromethane ≥98.0%

Supplier: TCI America

Synonyms: Fluorotribromomethane

T2304-5G
TCT2304-5GEA 107.8 USD
TCT2304-5G
Tribromofluoromethane ≥98.0%
Tribromofluoromethane

CAS Number: 353-54-8
MDL Number: MFCD00000366
Molecular Formula: CBr3F
Molecular Weight: 270.72
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 108
Melting point (°C): -73
Specific Gravity (20/20): 2.77


Other Product Names / Synonyms: Fluorotribromomethane

Formula: Br₃CF
MW: 270.72 g/mol
Boiling Pt: 108 °C
Melting Pt: –73 °C
Density: 2.77 (20 °C)
MDL Number: MFCD00000366
CAS Number: 353-54-8
https://us.vwr.com/stibo/web/std.lang.all/60/85/28996085.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR