2-Thiophenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Thiophene-2-sulphonyl chlorideThiophene-2-sulfonyl chloride

T2122-25G T2122-5G
TCT2122-25GEA 183 USD
TCT2122-25G TCT2122-5G
2-Thiophenesulfonyl chloride ≥98.0% (by GC, titration analysis)
2-Thiophenesulphonyl chloride

CAS Number: 16629-19-9
MDL Number: MFCD00005426
Molecular Formula: C4H3ClO2S2
Molecular Weight: 182.64
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 93
Melting point (°C): 33
Flash Point (°C): 113
Storage Temperature: 0-10°C

Formula: C₄H₃ClO₂S₂
MW: 182.65 g/mol
Boiling Pt: 93 °C
Melting Pt: 33 °C
Flash Pt: 113 °C
MDL Number: MFCD00005426
CAS Number: 16629-19-9
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/60/03/28996003.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR