3-Butynyl toluene-4-sulfonate ≥95.0%

Supplier: TCI America

Synonyms: 3-Butynyl 4-methylbenzenesulphonate3-Butynyl p-toluenesulfonate3-Butynyl p-toluenesulphonate3-Butynyl 4-methylbenzenesulfonate

T2046-5G
TCT2046-5GEA 149.21 USD
TCT2046-5G
3-Butynyl toluene-4-sulfonate ≥95.0%
3-Butynyl toluene-4-sulphonate

CAS Number: 23418-85-1
MDL Number: MFCD00041687
Molecular Formula: C11H12O3S
Molecular Weight: 224.27
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Color: Yellow
Specific Gravity (20/20): 1.19


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: p-Toluenesulfonic Acid 3-Butynyl Ester

Formula: C₁₁H₁₂O₃S
MW: 224.28 g/mol
Density: 1.19 (20 °C)
MDL Number: MFCD00041687
CAS Number: 23418-85-1
https://us.vwr.com/stibo/web/std.lang.all/59/73/28995973.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR