Tetrafluorohydroquinone ≥97.0%

Supplier: TCI America

Synonyms: 2,3,5,6-TetrafluorohydroquinoneTetrafluorobenzene-1,4-diol

T2003-5G
TCT2003-5GEA 57.25 USD
TCT2003-5G
Tetrafluorohydroquinone ≥97.0%
Tetrafluorohydroquinone

CAS Number: 771-63-1
MDL Number: MFCD00002340
Molecular Formula: C6H2F4O2
Molecular Weight: 182.07
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 175


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,2,4,5-Tetrafluoro-3,6-dihydroxybenzene

Formula: C₆H₂F₄O₂
Melting Pt: 175 °C
MDL Number: MFCD00002340
CAS Number: 771-63-1
https://us.vwr.com/stibo/web/std.lang.all/59/45/28995945.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR