2,4,6-Trifluorobenzonitrile ≥97.0%

Supplier: TCI America

T1908-5G
TCT1908-5GEA 105 USD
TCT1908-5G
2,4,6-Trifluorobenzonitrile ≥97.0%
2,4,6-Trifluorobenzonitrile

CAS Number: 96606-37-0
MDL Number: MFCD00042399
Molecular Formula: C7H2F3N
Molecular Weight: 157.10
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 61

Formula: C₇H₂F₃N
Melting Pt: 61 °C
MDL Number: MFCD00042399
CAS Number: 96606-37-0
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/58/66/28995866.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR