[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II) ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Cobalt tetramethoxyphenylporphyrinCobalt(II) meso-tetra(4-methoxyphenyl)porphine[5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphinato]cobalt(II)5,10,15,20-Tetrakis (4-methoxyphenyl)-21H,23H-porphine cobalt (II)

T1861-5G T1861-1G
TCT1861-1GEA 67.98 USD
TCT1861-1G TCT1861-5G
[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II) ≥96.0% (by titrimetric analysis)
[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II)

CAS Number: 28903-71-1
MDL Number: MFCD00010724
Molecular Formula: C48H36CoN4O4
Molecular Weight: 791.77
Purity/Analysis Method: 96.0% (T)
Form: Crystal
Color: Deep Blue
Lambda max.: 420 nm (THF)


Product Categories: Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials), Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: [5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphinato]cobalt(II)

Formula: C₄₈H₃₆CoN₄O₄
MW: 791.77 g/mol
MDL Number: MFCD00010724
CAS Number: 28903-71-1
https://us.vwr.com/stibo/web/std.lang.all/58/28/28995828.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR