TCIAT1762

T1762-5G
TCT1762-005GEA 168.11 USD
TCT1762-005G
TCIAT1762
CAS Number: 27761-02-0
MDL Number: MFCD00022505
Molecular Formula: C10H8O2S2
Molecular Weight: 224.29
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 109

Other Product Names / Synonyms: 2-Hydroxy-1,2-di-2-thienylethanone, alpha-Hydroxy-2-thenyl 2-Thienyl Ketone
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR