2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline ≥95.0%

Supplier: TCI America

T1529-5G T1529-1G
TCT1529-005GEA 155 USD
TCT1529-005G 76220-500
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline ≥95.0%
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline

CAS Number: 651-83-2
MDL Number: MFCD00091518
Molecular Formula: C7H2F7N
Molecular Weight: 233.09
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 186
Flash Point (°C): 87
Specific Gravity (20/20): 1.68


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Aminoheptafluorotoluene, 4-Amino-2,3,5,6-tetrafluorobenzotrifluoride

Formula: C₇H₂F₇N
Boiling Pt: 186 °C
Density: 1.68 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00091518
CAS Number: 651-83-2
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/55/42/28995542.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR