2-(Tribromomethyl)quinoline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
α,α,α-Tribromoquinaldine
T1453-5G
TCT1453-005GEA
78.12
USD
TCT1453-005G
2-(Tribromomethyl)quinoline ≥98.0% (by HPLC, titration analysis)
2-(Tribromomethyl)quinoline
CAS Number: 613-53-6
MDL Number: MFCD00006755
Molecular Formula: C10H6Br3N
Molecular Weight: 379.88
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 129
Other Product Names / Synonyms: alpha,alpha,alpha-Tribromoquinaldine
Formula:
C₁₀H₆Br₃N Melting Pt: 129 °C |
MDL Number:
MFCD00006755 CAS Number: 613-53-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...