1,2,3-Triphenylguanidine ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
T1316-25G
TCT1316-025GEA
139
USD
TCT1316-025G
1,2,3-Triphenylguanidine ≥97.0% (by HPLC, titration analysis)
1,2,3-Triphenylguanidine
CAS Number: 101-01-9
MDL Number: MFCD00020648
Molecular Formula: C19H17N3
Molecular Weight: 287.37
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 144
Formula:
C₁₉H₁₇N₃ Melting Pt: 144 °C |
MDL Number:
MFCD00020648 CAS Number: 101-01-9 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...