Thiophene-2-carboxamide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
2-Thiophenecarboxamide
T1252-5G
TCT1252-005GEA
70.04
USD
TCT1252-005G
Thiophene-2-carboxamide ≥98.0% (by total nitrogen basis)
Thiophene-2-carboxamide
CAS Number: 5813-89-8
MDL Number: MFCD00010425
Molecular Formula: C5H5NOS
Molecular Weight: 127.16
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 182
Formula:
C₅H₅NOS MW: 127.17 g/mol Melting Pt: 182 °C |
MDL Number:
MFCD00010425 CAS Number: 5813-89-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...