p-Toluenesulfonylacetonitrile ≥99.0%

Supplier: TCI America

Synonyms: [(4-Methylphenyl)sulphonyl]acetonitrile

T0846-5G T0846-25G
TCT0846-025GEA 178.64 USD
TCT0846-025G TCT0846-005G
p-Toluenesulfonylacetonitrile ≥99.0%
p-Toluenesulphonylacetonitrile

CAS Number: 5697-44-9
MDL Number: MFCD00039487
Molecular Formula: C9H9NO2S
Molecular Weight: 195.24
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 149


Other Product Names / Synonyms: p-(Cyanomethylsulfonyl)toluene, p-Tolylsulfonylacetonitrile

Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Melting Pt: 149 °C
MDL Number: MFCD00039487
CAS Number: 5697-44-9
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/49/55/28994955.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR