Bis(2,4-pentanedionato)tin(IV) dichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tin(IV) Acetylacetonate DichlorideTin(IV) Chloride Bis(2,4-pentanedionate)Acetylacetone Tin(IV) Dichloride Salt

T0650-25G
TCT0650-025GEA 395.61 USD
TCT0650-025G
Bis(2,4-pentanedionato)tin(IV) dichloride ≥98.0% (by titrimetric analysis)
Bis(2,4-pentanedionato)tin(IV) dichloride

CAS Number: 16919-46-3
MDL Number: MFCD00015316
Molecular Formula: C10H14Cl2O4Sn
Molecular Weight: 387.83
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 206


Other Product Names / Synonyms: Acetylacetone Tin(IV) Dichloride Salt, Tin(IV) Acetylacetonate Dichloride

Formula: C₁₀H₁₄Cl₂O₄Sn
MW: 387.83 g/mol
Melting Pt: 206 °C
MDL Number: MFCD00015316
CAS Number: 16919-46-3
https://us.vwr.com/stibo/web/std.lang.all/48/28/28994828.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR