7,7,8,8-Tetracyanoquinodimethane ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
1,4-Bis(dicyanomethylene)cyclohexadiene
T0078-25G
T0078-5G
TCT0078-025GEA
218.23
USD
TCT0078-025G
TCT0078-005G
7,7,8,8-Tetracyanoquinodimethane ≥98.0% (by HPLC, total nitrogen)
7,7,8,8-Tetracyanoquinodimethane
CAS Number: 1518-16-7
MDL Number: MFCD00011664
Molecular Formula: C12H4N4
Molecular Weight: 204.19
Purity/Analysis Method:
Form: Crystal
Color: Pale Yellow
Product Categories: Materials Chemistry, Organic Transistor Materials, Molecular Conductors
Other Product Names / Synonyms: 1,4-Bis(dicyanomethylene)cyclohexadiene, TCNQ
Formula:
C₁₂H₄N₄ MW: 204.19 g/mol |
MDL Number:
MFCD00011664 CAS Number: 1518-16-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...