Tetrabromobisphenol A ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3,3',5,5'-Tetrabromobisphenol A
CAS Number: 79-94-7
MDL Number: MFCD00013962
Molecular Formula: C15H12Br4O2
Molecular Weight: 543.88
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 316
Melting point (°C): 180
Product Categories: Materials Chemistry, Resin Additives, Monomers, Bisphenol A type Compounds (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 2,2-Bis(3,5-dibromo-4-hydroxyphenyl)propane, 4,4'-Isopropylidenebis(2,6-dibromophenol)
Formula:
C₁₅H₁₂Br₄O₂ Boiling Pt: 316 °C Melting Pt: 180 °C |
MDL Number:
MFCD00013962 CAS Number: 79-94-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...