Sulfadiazine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-N-(2-pyrimidinyl)benzenesulfonamideN-(2-Pyrimidinyl)sulfanilamide4-Amino-N-(2-pyrimidinyl)benzenesulphonamide

S0579-25G
TCS0579-25GEA 36 USD
TCS0579-25G
Sulfadiazine ≥99.0% (by HPLC, titration analysis)
Sulfadiazine

CAS Number: 68-35-9
MDL Number: MFCD00006065
Molecular Formula: C10H10N4O2S
Molecular Weight: 250.28
Purity/Analysis Method: 99.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 256
Storage Temperature: 0-10°C


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide, N-(2-Pyrimidinyl)sulfanilamide

Formula: C₁₀H₁₀N₄O₂S
MW: 250.28 g/mol
Melting Pt: 256 °C
MDL Number: MFCD00006065
CAS Number: 68-35-9
https://us.vwr.com/stibo/web/std.lang.all/41/92/28994192.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR