4,4'-Stilbenedicarboxylic acid ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,4-Stilbenedicarboxylic acid

S0557-25G
TCS0557-25GEA 60.9 USD
TCS0557-25G
4,4'-Stilbenedicarboxylic acid ≥95.0% (by HPLC)
4,4'-Stilbenedicarboxylic acid

CAS Number: 100-31-2
MDL Number: MFCD00013994
Molecular Formula: C16H12O4
Molecular Weight: 268.27
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Color: White

Formula: C₁₆H₁₂O₄
MW: 268.27 g/mol
MDL Number: MFCD00013994
CAS Number: 100-31-2
https://us.vwr.com/stibo/web/std.lang.all/41/86/28994186.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR