Sodium tetrachloropalladate(II) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Sodium palladium (II) chloride
, Disodium tetrachloropalladate
, Palladium sodium chloride
S0540-1G
TCS0540-1GEA
304.68
USD
TCS0540-1G
Sodium tetrachloropalladate(II) ≥98.0% (by titrimetric analysis)
Sodium tetrachloropalladate(II)
CAS Number: 13820-53-6
MDL Number: MFCD00003487
Molecular Formula: H2Cl4Pd
Molecular Weight: 294.20
Purity/Analysis Method:
Form: Crystal
Color: Reddish Yellow
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Palladium(II) Sodium Chloride
Formula:
Cl₄Na₂Pd MW: 294.21 g/mol |
MDL Number:
MFCD00003487 CAS Number: 13820-53-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...