Salbutamol sulfate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Albutarol sulfate
CAS Number: 51022-70-9
MDL Number: MFCD00055200
Molecular Formula: C13H21NO3
Molecular Weight: 288.35
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 180
Flash Point (°C): 250
Product Categories: Bioscience, Adrenoceptor Agent Ingredients
Other Product Names / Synonyms: 5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzyl Alcohol Hemisulfate, 2-(tert-Butylamino)-1-(4-hydroxy-3-hydroxymethylphenyl)ethanol Hemisulfate
Formula:
(C₁₃H₂₁NO₃)₂.H₂SO₄ MW: 576.702 g/mol Melting Pt: 180 °C Flash Pt: 250 °C |
MDL Number:
MFCD00055200 CAS Number: 51022-70-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...